missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Pipet Tips, 0.5 to 5mL
GREENER_CHOICE

Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene. Pipet Tips, 0.5 to 5mL allow pipetting from narrow volume measuring equipment. Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.

Brand:  BRAND™ 702605

 View more versions of this product

Product Code. 10135350

  • £99.35 / Pack of 28
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured using only high-purity polypropylene, free from oleamide and diHEMDA.
  • Slender shape allows pipetting even from narrow volume measuring equipment such as volumetric flasks with NS 12/21.
  • Suitable for Transferpette™ and Thermo Fisher Scientific FINNPIPETTE™ microliter pipettes.
  • Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
  • Autoclavable at 121°C (2bar). 

 


TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5 to 5 mL
Colorless
16 cm
TipBox
Transferpette™, Thermo Scientific™ Finnpipette™ Microliter Pipette
Non-sterile
Autoclavable
DiHEMDA- and oleamide-free
Slender
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers