Learn More
Gilson™ PIPETMAN™ Comfort Pipette Handles
For use with PIPETMAN Classic, Neo, G and Neo Multichannel pipettes
Brand:  Gilson™ F161486
Code : NEW
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 15394867
  • £57.75 / Pack of 3
In Stock
to see stock.
Add to basket
Description
Description
  • Simple to use and requires no tools
  • Designed to help reduce pipetting fatigue
  • Grip to support daily pipetting activity
  • Aids in reducing the risk of Repetitive Strain Injuries
  • Autoclavable at 121°C (252°F), 1 atm for 20 min.
Specifications
Specifications
Blue
Handle
PIPETMAN Classic, Neo, G and Neo Multichannel pipettes
3/Pk.