Learn More
Gilson™ PIPETMAN G 4-Pipette Kit
PIPETMAN G builds a new level of comfort into the Gilson™ pipette range with a redesigned piston that significantly reduces the force needed to aspirate and dispense liquids, a benefit during extended pipetting sessions which helps alleviate common hand stresses.
Brand: Gilson™
867.83 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 1.80000kg
Description
- Contains: 4 PIPETMAN G Pipettes (P2G, P20G, P200G, and P1000G), 3 racks of certified quality PIPETMAN DIAMOND Tips (D10L, D200 and D1000), 4 SINGLE™ pipette holders, and 1 Gilson Guide to Pipetting
- Maintains the trusted durability, accuracy and reliability of the PIPETMAN Classic
- Requires some of the lowest pipetting forces to operate (3 newetons on average)
- Lifetime warranty
- Manual air displacement type
Specifications
Polypropylene | |
1 | |
Manual Air Displacement Pipette Kit |
Pipetman™ G | |
Pipetman™ G | |
0.2 to 2, 100 to 1000, 2 to 20, 20 to 200 μL |