Learn More
Gilson™ PIPETMAN™ L Multichannel Pipette
GREENER_CHOICE
Designed for performance and comfort, PIPETMAN™ L Multichannel Pipette comes in a comprehensive range of models, adapted to your needs in the lab. Comfortable, precise, accurate, and reliable, PIPETMAN™ L variable volume multichannel models offer optimized performance across channels for uniform sample delivery and are perfect solutions when pipetting on microplates.
£969.00 - £1120.00
Specifications
Compatible Tips Most major brands
Autoclavable Autoclavable
Color White/Blue
Dispensing Speed Manual
Disposable Reusable
View More Specs
Products 4
Product Code Brand Volume (Metric) Number of Channels Model Price Quantity & Availability  
Product Code Brand Volume (Metric) Number of Channels Model Price Quantity & Availability  
17349013
GREENER_CHOICE
View Documents Promotion Details
Gilson™
FA10035
200 μL 8 8x200LVR
£969.00
Each
In Stock
to see stock.
Add to basket
17369013
GREENER_CHOICE
View Documents Promotion Details
Gilson™
FA10037
300 μL 8 8x300LVR
£969.00
Each
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
17359013
GREENER_CHOICE
View Documents Promotion Details
Gilson™
FA10036
200 μL 12 12x200LVR
£1120.00
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
17379013
GREENER_CHOICE
View Documents Promotion Details
Gilson™
FA10038
300 μL 12 12x300LVR
£1120.00
Each
In Stock
to see stock.
Add to basket
Description
Description
Available as a fixed or variable volume, PIPETMAN™ L air-displacement pipettes cover a full volume range, including:
  • Variable volume single channel models covering a volume range from 0.2 μL to 10 mL
  • Variable volume multichannel models covering a volume range from 0.5 μL to 1200 μL, including four models equipped with V-ring (VR) at the bottom of the tip holders for a proper fit with most tip brands, covering a volume range from 20 μL to 300 μL
  • Fixed volume single channel models covering a volume range from 1 μL to 5000 μL
Specifications
Specifications
Most major brands
White/Blue
Reusable
Polypropylene
No
Pipette
Certified Quality
Autoclavable
Manual
Ergonomic
Pipetman L
Non-sterile
Manual Liquid Handling
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Gilson™ PIPETMAN™ L Multichannel Pipette