missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAX™

PIPETMAX™ is an automated pipetting solution for the efficient processing of high-throughput biological assays, all in a benchtop size that easily fits into any lab.

Brand:  Gilson™ 32100000

 View more versions of this product

Product Code. 16626165

  • £31685.00 / Each
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

PIPETMAX is built upon PIPETMAN™ technology, helping to reduce costly technical replicates and maximize reproducibility by eliminating inconsistencies due to common pipetting errors. The automated system frees up time devoted to tedious pipetting jobs that improve lab productivity and allows you get to results faster. Several pre-developed, validated protocols are available for download while custom protocols can be written to accommodate any workflow.

  • Hardware and software of the PIPETMAX are customizable to fit specific needs of the assay, allowing you to configure and customize runs to any reagent, kit, or protocol
  • 8-channel cassettes can be set to use 1 to 8 tips
  • With a volume range from 1 to 1000 μL, PIPETMAX is an ideal solution for qPCR, NGS reactions, and cell-based assays
  • Small footprint and easily fits on lab bench or under a fume hood
  • Easy to move to location most suited for the assays at hand
Specifications

Specifications

Automated Pipetter
8
1 to 1000 μL
96- and 384-well microplates, strip plates, and cell culture formats
Touchscreen tablet, or PC control via USB and TRILUTION™ micro software
Polypropylene
100/240 V
6.25 A
ISO 9001
Polypropylene, Glass
Liquid Handling
Standard Cover
TRILUTION™
50/60 Hz, 100/240 V AC, 24 V DC, 6.25 A, 150 W
150 W
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilson™ PIPETMAX™ > PIPETMAX™; Includes: Standard Cover

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.