Learn More
Eppendorf™ Pipette Holder 2 for one Xplorer™ Xplorer™ plus Pipette
Holder designed for wall mounting or for installation in Pipette Carousel 2
Brand: Eppendorf™ 3116000120
39.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Carries one Eppendorf Xplorer™ or Xplorer™ plus
- Same as Charger Shell 2 for Eppendorf Xplorer or Xplorer plus but without charging functionality
- Designed for wall mounting or for installation in Pipette Carousel 2
- Sticky tape for wall mounting included
Specifications
Sticky tape for wall mounting included |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
12Hours
8Minutes
29Seconds
10 -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
15Minutes
30Seconds
49 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
29Hours
8Minutes
29Seconds
10