Learn More
Eppendorf™ Pipette Manager - Connected Electronic Pipette System
Control Eppendorf's connected electronic pipettes via a dedicated WiFi for faster, more reproducible results. Remotely set up pipettes, receive pipetting technique guidance, digitally document activities, and more.
£81.20 - £975.00
Specifications
For Use With | Xplorer Electronic Pipette |
---|---|
For Use With (Equipment) | Eppendorf Xplorer™, Xplorer plus and Xplorer plus Move It™ Electronic Pipettes |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
17152079
|
Eppendorf™
1004000001 |
Control Panel for Electronic Pipettes |
£975.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17162079
|
Eppendorf™
4861000970 |
WiFi Model Including Battery |
£81.20
Each |
In Stock
Log in to see stock. |
|||||
Description
Enhance your pipetting routines with the Eppendorf Pipette Manager. Set volumes quickly via touch screen, benefit from predefined pipette settings for volatile, viscous and other challenging liquids and digitally document your pipetting activities. The standalone plug'n'play system connects up to 30 Eppendorf Xplorer™, Xplorer plus or Move It™ electronic pipettes (when upgraded with WiFi module) and tablets.
- Connected electronic pipette system to remotely set up electronic pipettes, ease handling of challenging liquids and digitally document pipetting steps
- Easily enter pipetting volumes and other parameters using the touch screen. Settings are immediately transferred to connected electronic pipettes, allowing you to start working right away
- The software-embedded Liquid Manager sets optimal pipetting settings (e.g., pre-wetting steps, speed) when working with viscous, volatile and other challenging liquid types
- Choose from predefined Eppendorf liquid types (e.g., ethanol or enzymes/antibodies) or create your own
- Automatically document pipetting activities via Pipetting Records and easily export your recordings as comprehensive PDF document via USB
- Manage your pipette fleet more effectively and enhance collaboration with integrated features like Pipette Set Creation or Liquid Type Sharing
- All Eppendorf Xplorer™, Xplorer plus and Xplorer plus Move It™ electronic pipettes are compatible (when upgraded with the Eppendorf Xplorer connect WiFi module)
- Gain additional pipetting modes for your Xplorer pipettes (Sequential Dispensing, Sequential Pipetting, Diluting, Reverse Pipetting, Multi Aspiration)
- Easily update your Xplorer, Xplorer plus and Xplorer plus Move It pipette software with the Pipette Manager to never miss the latest features
- The graphical user interface is based on the VisioNize™ touch interface concept that has been extensively tested by customers to reduce learning and adjustment time
Specifications
Xplorer Electronic Pipette | |
Eppendorf Xplorer™, Xplorer plus and Xplorer plus Move It™ Electronic Pipettes |