Learn More
Miele Professional™ Mobile Unit E 404/1
Mobile unit E 404/1 for the optimum loading of up to 38 pipettes
Brand:  Miele Professional™ 6727910
Product Code. 10717831
  • £2450.00 / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Suitable for 38 pipettes in 3 rows
  • 1st Row: 10 pipettes x 100 ml (up to 550 mm), bar distance 20 mm
  • 2nd Row: 14 pipettes x 25 ml, bar distance 26 mm
  • 3rd Row: 14 pipettes x 10 ml, bar distance 26 mm
  • Without bracket for magnetic strip for mobile unit recognition
Specifications
Specifications
Holds 38 pipettes
1Each
E 404/1 Mobile injector
Pipettes