Learn More
Heathrow Scientific™ Pipette Starter Kit
The Pipette Starter Kit is convenient for new labs or for customers who want a new set of pipettes covering the most commonly used volumes. The kit includes 3 single channel Pearl™ Pipettes, a Universal Carousel Pipette Stand and five Reagent Reservoirs.
Brand: Heathrow Scientific™ 120554
398.35 GBP valid until 2024-12-31
Use promo code "21904" to get your promotional price.
Additional Details : Weight : 7.00000kg
Description
Universal Carousel Pipette Stand (120499):
- Holds up to eight single-channel, four multi-channel, or a combo of both pipettes
- Weighted base allows for easy removal or replacement of pipettes, with one hand
- Pipettes are held in a vertical position to help prevent contamination
- Conveniently rotate stand head to select or store pipettes
- Quick grip handle to move pipette stand (loaded or unloaded)
- Assorted color options allow for color coding pipette location
- The unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants
Pearl™ Pipettes (HS120104 100-1000 μL, HS120102 20-200 μL, and HS120099 2-20 μL)
- Internal metal parts for increased accuracy and calibration stability, and long life
- Easy volume adjustment with locking tab to prevent accidental change
- Separate tip ejector
- Large digital volume display
- Labeled with volume range
- Both top and side have colored coded components for quick visual confirmation
- CE certified
- Autoclavable
Reagent Reservoirs: (120644):
- Tapered reservoir bottoms form into a narrow trough to aid full sample recovery
- Disposable
- Sterile
Specifications
Autoclavable | |
Single-use | |
ABS Stand | |
Sterile | |
Pearl Pipette: Digital | |
Certified Quality |
Assorted | |
Fingerhook | |
1 | |
Pipette Kit | |
3 Single Channel Pearl™ Pipettes, Universal Carousel Pipette Stand, 5 Reagent Reservoirs |