missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Pipette Tip Boxes Zero Waste Box™

Recycle Pipette Tip Boxes using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
£105.75 - £191.81
Specifications
Color | Beige, Green, Red, White |
---|---|
Disposable | Single-use |
Latex Free | Yes |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17815725
![]() |
Terracycle™
CS180-S-UK |
Small |
£105.75
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
17855885
![]() |
Terracycle™
CS180-M-UK |
Medium |
£140.40
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
17865905
![]() |
Terracycle™
CS180-L-UK |
Large |
£191.81
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
Description
Use this Zero Waste Box to recycle:
- Empty pipette tip boxes
Do not send in:
- Pipette tips, tubes and rigid lab plastics that are contaminated with materials considered hazardous or dangerous
- Autoclaved material
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass and medical waste
Important:
- We do not collect any hazardous waste containing substances with toxic, reactive, corrosive, infectious, flammable, explosive properties or harmful to human health and the environment.
- Please make sure that you do not mix hazardous waste with non-hazardous waste.
Specifications
Beige, Green, Red, White | |
Yes | |
Not Autoclavable | |
Recycling Box |
Single-use | |
Non-sterile | |
Cardboard, Plastic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Terracycle™ Pipette Tip Boxes Zero Waste Box™