Learn More
Heathrow Scientific Pipette Tip Disposal Box, Clear
The Pipette Tip Disposal Box's quick flip of port cover makes tip ejection easy. It is convenient for one-handed disposal of used pipette tips and eliminates any need to handle pipette tips used with hazardous fluids.
Brand: Heathrow Scientific HS234634
This item is not returnable.
View return policy
Description
- The 3/8 in. (10 mm) thick acrylic provides protection from Beta radiation
- The large, hinged lid allows easy disposal of contents
- Use in conjunction with small biohazard disposal bags, if hazardous fluids are not acrylic friendly
Specifications
No | |
Non-sterile | |
Reusable |
Pipette Tip Disposal Box | |
Pipette Tips |