missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Pipette Workstation
GREENER_CHOICE

Organize your pipettes and tips in the Pipette Workstation and keep pipettes and disposable tips together in one convenient stand.

Brand:  Heathrow Scientific™ HS234631

Product Code. 16340746

  • £159.04 / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Acrylic workstation is opaque on sides with clear front and bin lids for easy content identification
  • Holds up to four pipettes on top shelf
  • Dual bins hold multiple size tips
  • Self-closing lid provides dust-free storage
  • Oversized lip allows for easy access when wearing gloves
Specifications

Specifications

Pipette Workstation
Pipettes
Acrylic
9.6 in.
6.5 in.
9.7 in.
Clear
4
Pipettes
24.4 cm
16.5 cm
24.6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heathrow Scientific™ Pipette Workstation > Pipette Workstation; Channels: 4; Color: Clear

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.