missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566524
This item is not returnable.
View return policy
Description
Immunogen sequence: NLNNLSSPSLN SAVPTPACPY APPTPPY Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Pilx2 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. This protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression. It plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes, is involved in the development of the eye, tooth and abdominal organs, and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. Mutations in this protein are associated with Axenfeld-Rieger syndrome, iridogoniodysgenesis syndrome, and sporadic cases of Peters anomaly. A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development.Specifications
PITX2 | |
Polyclonal | |
Unconjugated | |
PITX2 | |
9430085M16Rik; all1-responsive gene 1; ALL1-responsive protein ARP1; Arp1; Brx1; BRX1 homeoprotein; Brx1b; Homeobox protein PITX2; homeodomain transcription factor 2; IDG2; IGDS; IGDS2; IHG2; IRID2; Munc30; orthodenticle-like homeobox 2; Otlx2; paired like homeodomain 2; paired like homeodomain transcription factor 2; paired-like homeodomain 1; paired-like homeodomain 2; paired-like homeodomain transcription factor 1; paired-like homeodomain transcription factor 2; paired-like homeodomain transcription factor 2c; paired-like homeodomain transcription factor Munc 30; Pituitary homeobox 2; PITX1; Pitx2; Pitx-2; Pitx2c; Ptx2; Rgs; Rieg; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; RIEG1; rPtx2; RS; S a 1 glycoprotein; S a1glycoprotein; S alpha 1 glycoprotein; S a 1 glycoprotein; S a1glycoprotein; Solurshin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5308 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99697 | |
PITX2 | |
Recombinant Human PITX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only