missing translation for 'onlineSavingsMsg'
Learn More

PKC Sampler Kit, BD

Brand:  BD Biosciences 611421

Additional Details : Weight : 0.13200kg

Product Code. 15825929

  • £1835.00 / 1 set
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

Host Species:
Isotype: IgG1, IgG2a, IgG2b, IgG1, IgM
Cross Reactivity Species: Hu, Rt

Western Blotting

Specifications

Specifications

Unconjugated
Affinity Purified
10 μg (Each)
IgG1, IgG2a, IgG2b, IgG1, IgM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers