missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKD2L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13117-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. This protein functions as a calcium-regulated nonselective cation channel. Alternative splice variants have been described but their full length sequences have not been determined.Specifications
PKD2L1 | |
Polyclonal | |
Unconjugated | |
PKD2L1 | |
PCL, PKD2L, PKD2L1, PKDL, Polycystin 2 homolog, Polycystin 2L1, Polycystin L, Polycystin L1, TRPP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
293937, 329064, 9033 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2A259, Q9P0L9 | |
Pkd2l1 | |
PKD2L1 Fusion Protein Ag3807 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |