missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ PKI [14 22] Myristoylated PTD
PKI [14 22] Myristoylated PTD
Brand: Novex™ 77409
Code : 50
Additional Details : Weight : 0.50000kg
Description
cAMP-dependent protein kinase (PKA) inhibitor. This peptide is a highly specific inhibitor of cAMP-dependent protein kinase (PKA).Format/Formulation: Lyophilized trifluoroacetate salt, >97% purity by HPLC.
Molecular Weight: 1208
Specifications
Store in freezer (-5 to -30°C). | |
HPLC | |
PKA | |
cAMP-Dependent Protein Kinase (PKA) Inhibitor | |
1209 | |
Lyophilized |
5566, 5567, 5568 | |
Dry Ice | |
Non-sterile | |
Protein | |
Kinase Inhibitor, Kinase Inhibitor (PKA Inhibitor) |
For Research Use Only. Not for use in diagnostic procedures.