missing translation for 'onlineSavingsMsg'
Learn More

PKM2 Mouse anti-Human, Mouse, Rat, Clone: 1C11C7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60268-1-IG-150UL

 View more versions of this product

Product Code. 16837325

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

PKM2, Pyruvate kinase isozymes M1/M2, has 2 isoforms. The immunogen of this antibody is M2 isoform, also called PKM2.

PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.
Specifications

Specifications

PKM2
Monoclonal
0.53 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11980, P14618, P52480
PKM
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
1C11C7
Unconjugated
PKM
CTHBP, OIP 3, OIP3, Opa interacting protein 3, p58, PK2, PK3, PKM, PKM1, PKM2, PKM2 pyruvate kinase, muscle, Pyruvate kinase 2/3, Pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, TCB, THBP1, Tumor M2 PK
Mouse
Protein G
RUO
18746, 25630, 5315
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers