Learn More
Fisherbrand™ Plain Glass Plates with Bonded Spacers
Product Code. 11867643 Shop All Fisherbrand Products
Click to view available options
Includes:
0.5mm bonded spacers
0.75mm bonded spacers
1.5mm Bonded Spacers
2.0mm Bonded Spacers
Unit Size:
Pack of 2
Product Code. 11867643
Only null left
Add to basket
Add to basket
Accessory for use with OmniPAGE mini vertical Electrophoresis systems.
  • Accessory for use with OmniPAGE mini vertical Electrophoresis systems
Specifications
Type Plain Glass Plate
Gel Dimensions 100 x 100 mm
Includes 0.75mm bonded spacers