missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Plain PTFE Stir Bars
Provides stable stirring action. Fisherbrand™ Plain PTFE Stir Bars provide greater turbulence than cylindrical stir bars.
£9.18 - £44.90
Specifications
Autoclavable | Yes |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Color | White |
Description | PTFE encapsulated Alnico V magnet core |
For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
16215781
|
Fisherbrand™
|
20 mm | 7 mm | 10/Pk. |
£13.57
Pack of 10 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
16275781
|
Fisherbrand™
|
25 mm | 6 mm | 10/Pk. |
£14.34
Pack of 10 |
In Stock
Log in to see stock. |
|||
16285771
|
Fisherbrand™
|
12 mm | 4.5 mm | 10/Pk. |
£9.18
Pack of 10 |
In Stock
Log in to see stock. |
|||
11878962
|
Fisherbrand™
|
75 mm | 13 mm | 2/Pk. |
£30.34
Pack of 5 |
In Stock
Log in to see stock. |
|||
11808962
|
Fisherbrand™
|
13 mm | 8 mm | 10/Pk. |
£15.45
Pack of 10 |
In Stock
Log in to see stock. |
|||
11828962
|
Fisherbrand™
|
25 mm | 10 mm | 5/Pk. |
£24.66
Pack of 10 |
In Stock
Log in to see stock. |
|||
16235791
|
Fisherbrand™
|
40 mm | 8 mm | 5/Pk. |
£20.33
Pack of 10 |
In Stock
Log in to see stock. |
|||
16285781
|
Fisherbrand™
|
25 mm | 8 mm | 5/Pk. |
£17.80
Pack of 10 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
11858962
|
Fisherbrand™
|
65 mm | 13 mm | 2/Pk. |
£33.95
Pack of 5 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
10306280
|
Fisherbrand™
|
50 mm | 7 mm | 2/Pk. |
£26.37
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
16295771
|
Fisherbrand™
|
15 mm | 4.5 mm | 10/Pk. |
£9.95
Pack of 10 |
In Stock
Log in to see stock. |
|||
16295791
|
Fisherbrand™
|
50 mm | 8 mm | 2/Pk. |
£23.80
Pack of 10 |
In Stock
Log in to see stock. |
|||
11838962
|
Fisherbrand™
|
35 mm | 10 mm | 5/Pk. |
£42.31
Pack of 10 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
16205801
|
Fisherbrand™
|
50 mm | 10 mm | 2/Pk. |
£36.35
Pack of 10 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
10010110
|
Fisherbrand™
|
60 mm | 7 mm | 2/Pk. |
£32.20
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
16205781
|
Fisherbrand™
|
20 mm | 6 mm | 10/Pk. |
£13.57
Pack of 10 |
In Stock
Log in to see stock. |
|||
16295781
|
Fisherbrand™
|
30 mm | 6 mm | 5/Pk. |
£17.08
Pack of 10 |
In Stock
Log in to see stock. |
|||
16255801
|
Fisherbrand™
|
70 mm | 10 mm | 2/Pk. |
£44.90
Pack of 10 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||
16215801
|
Fisherbrand™
|
60 mm | 10 mm | 2/Pk. |
£41.35
Pack of 10 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||
11818952
|
Fisherbrand™
|
35 mm | 7 mm | 5/Pk. |
£18.90
Pack of 10 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
16265801
|
Fisherbrand™
|
80 mm | 10 mm | 2/Pk. |
£25.50
Pack of 5 |
In Stock
Log in to see stock. |
|||
16225801
|
Fisherbrand™
|
60 mm | 8 mm | 2/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16225791
|
Fisherbrand™
|
35 mm | 6 mm | 5/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11848962
|
Fisherbrand™
|
50 mm | 10 mm | 2/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Stable, general-purpose magnetic stir bar
- Provides greater turbulence than cylindrical stir bars
- Autoclavable
Specifications
Yes | |
White | |
Containers & Vessels / Stirrer Hot Plates |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Alnico V magnet core |