missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human lys-Plasmin Native Protein

Product Code. 11574151
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
This item is not returnable. View return policy

Product Code. 11574151

Brand: Invitrogen™ RP43124

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

RP-43124 contains native human Plasmin protein. RP-43124 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00747
Concentration 5.7 mg/mL
For Use With (Application) Neutralization
Formulation 50% water with 50% glycerol and no preservative
Gene ID (Entrez) 5340
Molecular Weight (g/mol) 83 kDa
Name Human lys-Plasmin
Purification Method SDS-PAGE
Quantity 500 μg
Source Human
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activation peptide; Angiostatin; plasmin; Plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; Plasminogen; PLG
Common Name lys-Plasmin
Gene Symbol PLG
Product Type Protein
Conjugate Unconjugated
Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.