Learn More
Fisherbrand™ Plastic Beaker with Cover
A safe and economical choice for ultrasonic cleaning applications. Fisherbrand™ Plastic Beaker with Cover is for use with S10 and S10-H benchtop ultrasonic cleaners.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 1.00000kg
Description
- Graduated
- Sturdy and durable
Specifications
Lid | |
Ultrasonic Bath, Elma S10 H | |
Plastic |