missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Plastic Bio-Hazard Trays
Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
460 x 260 mm
Unit Size:
Each
Description
- Available in general purpose, biohazard and radiation safety formats
- Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
- All trays supplied with one free liner when purchased
- Additional environmentally friendly APET liners are provided in packs of 25
Specifications
Specifications
| Dimensions (L x W) | 460 x 260 mm |
| Color | White |
| Material | Plastic |
| Type | Spill Containment Tray |
| For Use With | Safe containment of spillages, lab bench protection |
| Includes | Liner |
| No. per Pack | 1 Each |
| Shape | Rectangular |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction