Learn More
Lovibond™ Plastic cell
For Comparator 2000+ and CHECKIT comparators
Brand: Lovibond™ 145505
This item is not returnable.
View return policy
Description
For Comparator 2000+ and CHECKIT comparators
Plastic cell, 13.5 mm path length, with grey lid, for Comparator 2000+ and CHECKIT comparators.
Specifications
Set of 5 | |
13.5 mm pathlength | |
Plastic Cell |