missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plastic Drum, 300L, for HyPerforma™ Single-Use Mixer DS 300
Brand: HyPerforma SH30959.04
Additional Details : Weight : 1.00000kg
Specifications
60 to 300 L | |
HyPerforma™ | |
DS 300 Single-Use Mixer |
61 x 122 cm (24 x 48 in.) | |
Fewer resources used and less waste |