Learn More
Sperian™ Honeywell™ Plastic RD40 Filters
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Plastic RD40 Filters are positioned low to prevent obstruction in the field of vision.
Brand: Sperian™ 1786000
Code : NEW
Additional Details : Weight : 0.12600kg
Description
- Body made of plastic provides optimum resistance
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Filter | |
RD40 | |
Plastic Filters | |
Full-Face Mask And Half-Mask Equipped With An Rd40 Connector |
E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 0070-003-079-06-98-0039, EC Attestation- EC Attestation, EC Attestation Number- 0070-003-079-06-98-0039, Certifications- EN 14387, EN 143, EN 12; EN 143 : 1998 type P3 | |
ABS Material With Plastic Cover | |
Dust and particulates protection |