Learn More
Sperian™ Honeywell™ Plastic RD40 Filters
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Plastic RD40 Filters are positioned low to prevent obstruction in the field of vision.
Brand: Sperian™ 1788155
Code : NEW
Additional Details : Weight : 0.38700kg
Description
- Body made of plastic provides optimum resistance
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Filter | |
RD40 | |
Plastic Filters |
E.C. Declaration of Conformity- EC Category PPE- 3, EC Certificate Number- 7793 A/13/8 PSA, EC Attestation- EC Attestation, EC Attestation Number- 7793 A/13/8 PSA, Certifications- EN 14387, Declaration of conformity DEKRA - 1788155 |
|
ABS material with plastic cover - Activated and/or impregnated charcoal | |
Gas vapor and particulates protection - A2B2E2K2 P3 large capacity for long duration work |