Learn More
Sperian™ Honeywell™ Plastic RD40 Filters
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Plastic RD40 Filters are positioned low to prevent obstruction in the field of vision.
Brand: Sperian™ 1788161
Code : NEW
Additional Details : Weight : 0.28800kg
Description
- Body made of plastic provides optimum resistance
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Filter | |
RD40 | |
Plastic Filters | |
Full-Face Mask Equipped With An RD40 Connector |
E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 0070-236-079-03-06-0006, EC Attestation- EC Attestation, EC Attestation Number- 0070-236-079-03-06-0006 | |
ABS material with plastic cover - High activated charcoal | |
Gas/vapor and particulates protection - AX large capacity for long duration work |