Learn More
Nickel Electro™ Plastic Flat Lid
Reduce the evaporation and noise in an Ultrasonic Bath. Nickel Electro™ Clifton™ Plastic Flat Lid only requires one piece construction.
Brand: Nickel Electro™
Code : 03
Additional Details : Weight : 0.50000kg
Description
- One piece construction
- Reduces evaporation
- Reduces noise
- For use with Ultrasonic Bath
Specifications
Ultrasonic Bath SW12H | |
Plastic |