Learn More
Cole-Parmer™ Indenti-plug™ Plastic Foam Stopper
Product Code. p-8000137 Shop All Cole-Parmer Products
Click to view available options
Diameter (Metric) Inner:
35 to 45 mm
45 to 50 mm
Unit Size:
Pack of 100
Pack of 80
Product Code. 11735289
Brand: Cole-Parmer™ 0629880
Only null left
Add to basket
Add to basket
Keep the contents of your containers secure with these durable and reusable stoppers. Indenti-plug™ Plastic Foam Stoppers can also be used as a cleaner for pipettes, tubes, vials and precision tubing.
  • Breathable, nonabsorbent, chemical resistant and heat tolerant
  • Open-cell foam has a uniform density
  • Leaves no fibers
  • Autoclavable
Specifications
Diameter (Metric) Inner 45 to 50 mm
Quantity 80/Pk.
Autoclavable Autoclavable
Color White
Material Plastic
Product Type Plug Stopper