missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heinz Herenz Plastic Forceps
Plastic forceps EO sterile, PS.
Brand: Heinz Herenz 2000003
This item is not returnable.
View return policy
Description
- Tweezers come extremely handy for several tasks, including picking up tiny bits that are difficult for fingers to easily grasp.
- Lab personnel or medical staff can use them for moving sterile items from place to place.
Specifications
Pointed | |
Polystyrene | |
Forceps | |
Sterile |
0.41 ft. | |
123.7 mm | |
Forceps | |
Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heinz Herenz Plastic Forceps > Standard Tip
Spot an opportunity for improvement?Share a Content Correction