Learn More
DWK Life Sciences Plastic Hose Connection, with silicone seal (VMQ), PP complete with screw cap PBT, straight
For safe connection of hoses.
Brand:  DWK Life Sciences 286221806
Product Code. 15804628
  • £327.00 / Pack of 100
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PP are used for lower thermal and chemical resistance requirements.
  • Hose olives enable the connection of hoses to laboratory apparatuses
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100/Pk.
8.6 mm