Learn More
Microspec™ Plastic Inoculation Loops
Available as aseptic manufactured or ethylene oxide sterilized.
Brand:  Microspec™ B5
 View more versions of this product
Product Code. 16634822
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Includes 250 Plastic Inoculation Loops
  • Aseptic Manufactured or Ethylene Oxide Sterilized
  • 200 mm length
Specifications
Specifications
Single-use
Non-sterile
200 mm
Microseal Pouch
Not Autoclavable
5 μL
Blue
250
Yes