Learn More
Microspec™ Plastic Inoculation Loops
Available as aseptic manufactured or ethylene oxide sterilized.
Brand:  Microspec™ B5G
 View more versions of this product
Product Code. 16644822
  • £34.25 / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Includes 250 Plastic Inoculation Loops
  • Aseptic Manufactured or Ethylene Oxide Sterilized
  • 200 mm length
Specifications
Specifications
Single-use
Sterile
200 mm
Microseal Pouch
5 μL
Blue
250
Not Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Microspec™ Plastic Inoculation Loops > 5 μL, Sterile