Learn More
Buerkle™ Plastic Powder Funnels
Powder funnel for safe filling of containers. 9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).
Brand:  Buerkle™ 9606-0240
14.36 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10085091
  • £19.60 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of PP/PE
  • For safe filling of powders
Specifications
Specifications
253 mm
50 mm
Polyethylene
For Foodstuffs