Learn More
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock
HPLC certified, non-sterile, two-part disposable syringes made of solvent robust polypropylene.
£37.11 - £76.77
Specifications
Material Polypropylene
Connection Type Luer Lock
View More Specs
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
12921031
View Documents
Fisherbrand™
10 36 2401
10 mL
£62.60
Pack of 100
12941031
View Documents
Fisherbrand™
20 36 2402
20 mL
£76.77
Pack of 100
12901031
View Documents
Fisherbrand™
05 36 2400
5 mL
£48.60
Pack of 100
12981021
View Documents
Fisherbrand™
02 36 2399
2 mL
£37.11
Pack of 100
Description
Description
  • Plastic Disposable Syringe
  • Polypropylene
  • Luer Lock
  • Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
  • No silicone or oil lubricant is required in the barrel
Specifications
Specifications
Polypropylene
Luer Lock
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock