Learn More
Fisherbrand™ Plastic PP Syringes, Luer Slip
HPLC certified, non sterile disposable syringes made of solvent robust polypropylene
Brand: Fisherbrand™ 10 36 2405
28.85 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- Plastic Disposable Syringe
- Polypropylene
- Luer Slip
- Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
- No silicone or oil lubricant is required in the barrel

Specifications
Polypropylene | |
10 mL | |
Leur Slip |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.