Learn More
Swann-Morton™ Plastic Scalpel Blade Remover, Sterile
Remove used blades from a scalpel with the Swann Morton™ Plastic Scalpel Blade Remover, Sterile. The blade is retained in the remover to ensure that they are disposed together.
Brand:  Swann-Morton™ 5502
Product Code. 12932575
  • £54.45 / Pack of 50
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sterile
  • Single use
  • Blade retained in the remover and both are disposed of together
Specifications
Specifications
Scalpel Blade Remover
Plastic
Scalpel
50