Learn More
Micronova™ Plastic Squeegee
These durably constructed squeegees are made with a polypropylene frame and a choice of either solid rubber or double foam blades for use in wash and production areas. Designed with an ACME threaded handle connection for use with most threaded plastic handles.
Brand: Micronova™ SQDB-24
Code : 03
Additional Details : Weight : 1.00000kg
Description
The Micronova Plastic Squeegees easily clean smooth or uneven surfaces and minimize contamination areas by reducing dirt and bacteria growth. Designed for use with STAH or STAEH series plastic handles. Available in a variety of colors.
- Durably constructed polypropylene squeegees
- Choice of solid rubber or double foam blades
- Excellent for use on smooth surfaces and uneven floor surfaces
- Prevents hazardous sparks when cleaning-up flammable fluids
- Available in 18 and 24 inch (45 and 60 cm) widths
- Use with STAH or STAEH handles for a secure fit
- Will not rust
Specifications
Squeegee, ACME Threaded End | |
Store in cool dry area when not in use. | |
No | |
STA Series Plastic Handles | |
1.5 in. |
Blue | |
61 x 3.8 cm (24 x 1.5 in.) | |
Squeegee for Floors, Walls and Windows | |
Foam Blade, Polypropylene frame | |
3.8 cm |