missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Plastic Sweatband
3M™ Plastic Sweatband suitable for use with 3M™ hard hats and headgear combinations.
£36.70 - £735.00
Specifications
Certifications/Compliance | CE Approved |
---|---|
Color | Black |
Material | Plastic |
Product Type | Sweatband |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15729867
|
3M™
7000103789 |
3M Headgear Combinations G500 Series |
£36.70
Pack of 10 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17267206
|
3M™
7000108029 |
All 3M™ Hard Hats |
£735.00
Pack of 250 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
15739957
|
3M™
7000108029 |
All 3M™ Hard Hats |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Replacement plastic sweatband suitable for use with 3M™ Hard Hats and 3M™ G500 Headgear Combinations
- Quick and easy to install
Specifications
CE Approved | |
Black | |
Plastic | |
Sweatband |