missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ LaboPlast™ Plastic Splinter-Proof Titrating Burettes

Allows for precise dosing and exact fine titration with the press button or micro-screw and precise discharge through the discharge jet. Buerkle™ Titrating Burets with Shatter Protection has a plastic sheathing which increases resistance to breaki

Brand:  Buerkle™ 9695-3050

81.41 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11991501

  • £141.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Burette with splinter-proof sheathing
  • Adjusted to Ex
  • Burette made of borosilicate glass 3.3
  • Shatter-proof, resistant to chemicals
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
  • Parts in contact with medium: PE, PP, natural rubber, borosilicate glass
  • Fast acting burette with Schellbach stripes based on Dr. Schilling's design
  • Plastic sheathing which increases resistance to breaking and protects against splinters
  • Allows for precise dosing and exact fine titration with the press button or micro-screw and precise discharge through the discharge jet
Specifications

Specifications

50 mL
Plastic-coated Borosilicate Glass
Schellbach Stripes, Splitter-Proof Sheathing, Storage Bottle and Immersion Tube
Titrating Buret
92 cm
92 cm
0.1 mm
DIN
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.