missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Plastic Twist Ties
Twist around bags and other lightweight items to keep contents inside. Azpack™ Plastic Twist Ties include a single wire insert for tightening.
£34.80 - £69.85
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12350579
|
Azpack™
12350579 |
100 mm |
£34.80
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
12360579
|
Azpack™
12360579 |
150 mm | N/A | ||||||
Description
- Flexible wire insert
- White plastic coating
- Disposable
- Reusable
Specifications
Plastic | |
White | |
1000/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title