Learn More
Kartell™ Plastilab™ Conical Stoppers
Features tapered conical-shaped bottom to ensure sealing action.
Brand: Kartell™ 0124300
Description
- Made from Polyethylene for tight and secure seal
- Raised embossed easy-to-read sizes
- Color-coded
Specifications
DIN 12242 | |
NS 14/23 | |
12.2 mm | |
Tapered Stopper |
Conical bottom | |
Brown | |
Polyethylene | |
Pentagonal Grip |