Learn More
Kartell™ Plastilab™ Two-Tier Holder for Vertical Pipette Stand
Product Code. 10684232
Click to view available options
:
Holds: 18 Small and 10 Large Holes Places
Product Code. 10684232
Brand: Kartell™ 0026500
Only null left
Add to basket
Add to basket
Consists of two-tiered pipette holder only, base and support rod are not included.
Consists of two-tiered pipette holder only, base and support rod are not included.
  • Made from polypropylene
  • Boss head allows holder to be raised or lowered on support rod
Specifications
Product Type Pipet Holder
Material Polypropylene
Positions 18 Small and 10 Large Holes Places
Quantity 1 Each
Diameter (Metric) Outer 175 mm