Learn More
BD Plastipak™ Centric Luer-lock Hypodermic Three-Piece Syringe
Plastic graduated syringe.  BD Medical™ Plastipak™ Centric Luer-lock Hypodermic Three-Piece Syringe for injection of fluid.
Brand:  BD 309658
Code : 02
Additional Details : Weight : 1.26000kg
 View more versions of this product
Product Code. 11303040
  • £154.80 / Pack of 200
In Stock
to see stock.
Description
Description
  • Plastic syringe
  • Includes luer-lock
Specifications
Specifications
Autoclavable
Polypropylene
Yes
Centic Luer-lock Hypodermic 3-piece Syringe
0.1 mL
3 mL
Yes
NF EN 20594-1, ISO 594-1 de December 1993, ISO 594-2 de september 1998
10.12 mm
69 mm
Yes
200/Pk.
Centric Luer Lock
Sterile