Learn More
Eppendorf™ Plate Carriers for Buckets
Carriers designed for MTP, DWP, PCR plates. Available for high-capacity buckets and plate/tube buckets
Brand:  Eppendorf™ 5920729004
Code : NEW
Additional Details : Weight : 0.56000kg
 View more versions of this product
Product Code. 15684614
  • £286.00 / Each
In Stock
to see stock.
Description
Description
  • Number of tubes per adapter/per rotor: 7/28(MTP)\n2/8(DWP) or 6/24 (MTP)\n2/8 (DWP)
  • Set of 2 pcs.
Specifications
Specifications
Plate Carrier for High-capacity Bucket
for MTP, DWP, PCR plates
6/24 (MTP)\n2/8 (DWP)