missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plate Count Agar

Enumerate viable organisms in milk, water, food and dairy products using this non-selective medium.

Brand:  Thermo Scientific™ PO0158A

12.66 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11973612

  • £26.05 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.

This standard medium meets the formulation of APHA, AOAC and PHLS.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tested in accordance with CEN ISO/TS 11133-2.
2° to 10°C, away from light
United Kingdom
Agar
Flow Wrapped - Single Layer
Straw, clear
90mm Monoplate
Plate Count Agar
19.5g ± 1.0g
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4498709 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberPO0158A
Lot Number4496115 Certificate TypeCertificate of Analysis Date06/03/2025 Catalog NumberPO0158A
Lot Number4489444 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog NumberPO0158A
Lot Number4487820 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberPO0158A
Lot Number4487775 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberPO0158A
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Plate Count Agar >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.