Learn More
Kinematica™ ST-P13/320 Holder System for PT 1200 E
Stainless steel plate stand with anti-slip mat made of silicone, with height-adjustable holder for the drive and for fastening the base station.
Brand: Kinematica™ 11040007
This item is not returnable.
View return policy
Description
- Stainless steel plate stand with anti-slip silicone mat
- Drive unit height adjustable allowing for hands free work
- Vessel Holder
- Safety Positioning ring: safety feature to prevent uncontrolled adjustment of drive unit
Specifications
Plate Stand | |
Polytron™ PT 1200 E | |
ST-P 13/320 |
Stand for PT 1200E with height-adjustable holder for the drive unit and fastening the base station. | |
Silicone Anti-Slip Mat |
Product Content Correction
Product Title
Kinematica™ ST-P13/320 Holder System for PT 1200 E > For use with: Polytron PT 1200 E
Spot an opportunity for improvement?Share a Content Correction