Learn More
Gilson™ PLATEMASTER™ 220 μL
GREENER_CHOICE
PLATEMASTER™ 220 μL is an easy-to-use, accurate solution for high-throughput manual pipetting of 96- and 384-well microplates.
Brand:  Gilson™ F110762
16444.27 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 15843192
  • £18970.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The PLATEMASTER™ 96-channel design greatly reduces the number of pipetting steps necessary to fill a microplate when compared to using regular manual multichannel pipettes. Typically, filling 96-well plates takes less than 10 seconds, and 384-wells can be filled in less than a minute using only four pipetting steps.
  • Reduces risk of human error in high-throughput applications
  • High accuracy and precision levels can equal that of 8- or 12-channel pipettes while avoiding the risk of skipping or repeating wells
  • All enzymatic reactions and cell-based applications across wells can be started and stopped at the same time
  • Improved well-to-well and plate-to-plate reproducibility from 0.5 μL to 220 μL pipetting volumes
  • As intuitive as a PIPETMAN™ and does not require programming, long training, or power for use
  • Can be fully adapted to a variety of application needs
  • Compact and portable design easily integrates into lab, allowing it to fit on most benchtops, under hoods, or in cold rooms
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pipettor
96
2 to 220 μL
PLATEMASTER™ P220
0 to 100 mV, 1 V, 10 V
ISO 9001
Polypropylene
Liquid Handling
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ PLATEMASTER™ 220 μL > PLATEMASTER™ 220 μL; Certifications/Compliance: ISO 9001; Channels:96 Channel; Material: Polypropylene; Volume (Metric): 2 to 220 μL; Model: PLATEMASTER™ P220