Learn More
Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Brand: Thermo Scientific™ 436006
404.99 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 76
Additional Details : Weight : 1.44900kg
Description
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)
- Minimal leaching
- Withstands vigorous washing
- Coating with lower amounts of reagent may be possible
- Control of orientation
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Specifications
Covalent | |
Standard | |
Colorimetric | |
30 Cases | |
Microplate | |
96 | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
5 | |
Peptides |
No | |
Without Lid | |
8 x 12 | |
Clear | |
Polystyrene | |
100 μL | |
Covalent Binding | |
30 | |
Unspecified |
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.