Learn More
Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Brand: Thermo Scientific™ 436023
464.64 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.60000kg
Description
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)
- Minimal leaching
- Withstands vigorous washing
- Coating with lower amounts of reagent may be possible
- Control of orientation
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Specifications
Breakable Strips | |
30 Cases | |
Clear | |
Polystyrene | |
100 μL | |
Amine | |
Round, C-bottom | |
5 | |
Peptides |
Colorimetric | |
Covalent | |
Microplate | |
350 μL | |
Non-sterile | |
350 μL | |
30 | |
Unspecified |
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.